Learn More
Manutan Shackles, 8.5T SWL bow, galvanized alloy steel
Product Code. 12841679
Encompass_Preferred
Product Code. 12841679 Supplier Manutan Supplier No. A339942
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • Galvanised alloy steel
  • Dimensions : d - 25mm ; D - 28mm ; w - 43mm ; s - 85mm ; e - 56mm