Learn More
Manutan Shackles, 4.75T SWL bow, galvanized alloy steel
Product Code. 12841639
Encompass_Preferred
Product Code. 12841639 Supplier Manutan Supplier No. A339911
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • Galvanised alloy steel
  • Dimensions : d - 19mm ; D - 22mm ; w - 31mm ; s - 59mm ; e - 44mm