missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Shackles, 2.0T large D, high tensile steel
Description
Use to securely connect lifting device to the load to be lifted.
- Dee - Pin
- Max. weight : 8500kg
- SWL : 2.00T
- High tensile steel
- Dimensions : d - 19mm ; D - 22mm ; w - 38mm ; s - 72mm ; e - 44mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction