Learn More
Manutan Shackles, 1.5T SWL bow, galvanized alloy steel
Product Code. 12868978
Encompass_Preferred
Product Code. 12868978 Supplier Manutan Supplier No. A340007
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • Galvanised alloy steel
  • Dimensions : d - 11mm ; D - 13mm ; w - 18mm ; s - 37mm ; e - 28mm