missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3TC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558059
This item is not returnable.
View return policy
Description
Immunogen sequence: SVLSFLYDKK YLPHLAVASV QQHGIQSAQG MSLPIWQVHL VLQNTTKLLG FPSPGWGEVS ALACPMLRQA LAACEELADR STQRALCLIL SKVYLEHRSP DGAIHYLSQ Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.
This gene encodes a protein with two N-terminal Src homology 3 (SH3) domains and 10 tetratricopeptide repeat (TPR) motifs, and is a member of a small gene family. The gene product has been proposed to be an adapter or docking molecule. Mutations in this gene result in autosomal recessive Charcot-Marie-Tooth disease type 4C, a childhood-onset neurodegenerative disease characterized by demyelination of motor and sensory neurons.
Specifications
| SH3TC2 | |
| Polyclonal | |
| Unconjugated | |
| SH3TC2 | |
| CMT4C; D430044G18Rik; KIAA1985; MNMN; PP12494; RGD1309038; SH3 domain and tetratricopeptide repeat-containing protein 2; SH3 domain and tetratricopeptide repeats 2; SH3 domain and tetratricopeptide repeats-containing protein 2; SH3TC2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TF17 | |
| SH3TC2 | |
| Recombinant protein corresponding to Human SH3TC2. Recombinant protein control fragment (Product #RP-96927). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction