missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3TC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584311
This item is not returnable.
View return policy
Description
Immunogen sequence: AITFMTQAVE ASAIAGVRAI VDHLVALAWL HVLHGQSPVA LDILQSVRDA VVASEDQEGV IANMVAVALK RTGRTRQAAE SYYRALRVAR DLGQQRN Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 62%.
SH3TC1 is a protein coding gene.
Specifications
| SH3TC1 | |
| Polyclonal | |
| Unconjugated | |
| SH3TC1 | |
| SH3 domain and tetratricopeptide repeat-containing protein 1; SH3 domain and tetratricopeptide repeats 1; SH3 domain and tetratricopeptide repeats-containing protein 1; SH3TC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TE82 | |
| SH3TC1 | |
| Recombinant protein corresponding to Human SH3TC1. Recombinant protein control fragment (Product #RP-105054). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction