missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3RF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561737
This item is not returnable.
View return policy
Description
Immunogen sequence: LISSSDPRAA ARIGDLAHLS CAAPTQDVSS SAGSTPTAVP RAASVSGEQG TPPKVQLPLN VYLALYAYKP QKSDELE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Has E3 ubiquitin-protein ligase activity.
Specifications
| SH3RF3 | |
| Polyclonal | |
| Unconjugated | |
| SH3RF3 | |
| 4831416G18Rik; E3 ubiquitin-protein ligase SH3RF3; Igfbp5-ip; insulin-like growth factor binding protein 5 interacting protein; plenty of SH3s 2; plenty of SH3s-2; POSH2; SH3 domain containing ring finger 3; SH3 domain-containing RING finger protein 3; SH3 multiple domains 4; SH3 multiple domains protein 4; SH3MD4; Sh3rf3; Sh3rf31 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 344558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEJ3 | |
| SH3RF3 | |
| Recombinant protein corresponding to Human SH3RF3. Recombinant protein control fragment (Product #RP-99674). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction