missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PXD2A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | SH3PXD2A |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18341376
|
Novus Biologicals
NBP3-16808-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18364697
|
Novus Biologicals
NBP3-16808-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SH3PXD2A Polyclonal antibody specifically detects SH3PXD2A in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| SH3PXD2A | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| adaptor protein TKS5, SH3 and PX domains 2A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 806-1105 of human SH3PXD2A (NP_055446.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 9644 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title