missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals SH3PXD2A Antibody, Novus Biologicals™

Product Code. 18307868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18307868 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18307868 Supplier Novus Biologicals Supplier No. H00009644B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

SH3PXD2A Polyclonal antibody specifically detects SH3PXD2A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3PXD2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. AAI56573.1
Gene Alias adaptor protein TKS5, SH3 and PX domains 2A
Host Species Mouse
Immunogen SH3PXD2A (AAI56573.1, 1 a.a. - 1105 a.a.) full-length human protein. MLAYCVQDATVVDVEKRRNPSKHYVYIINVTWSDSTSQTIYRRYSKFFDLQMQLLDKFPIEGGQKDPKQRIIPFLPGKILFRRSHIRDVAVKRLKPIDEYCRALVRLPPHISQCDEVFRFFEARPEDVNPPKEDYGSSKRKSVWLSSWAESPKKDVTGADATAEPMILEQYVVVSNYKKQENSELSLQAGEVVDVIEKNESGWWFVSTSEEQGWVPATYLEAQNGTRDDSDINTSKTGEEEKYVTVQPYTSQSKDEIGFEKGVTVEVIRKNLEGWWYIRYLGKEGWAPASYLKKAKDDLPTRKKNLAGPVEIIGNIMEISNLLNKKASGDKETPPAEGEGHEAPIAKKEISLPILCNASNGSAVGVPDRTVSRLAQGSPAVARIAPQRAQISSPNLRTRPPPRRESSLGFQLPKPPEPPSVEVEYYTIAEFQSCISDGISFRGGQKAEVIDKNSGGWWYVQIGEKEGWAPASYIDKRKKPNLSRRTSTLTRPKVPPPAPPSKPKEAEEGPTGASESQDSPRKLKYEEPEYDIPAFGFDSEPELSEEPVEDRASGERRPAQPHRPSPASSLQRARFKVGESSEDVALEEETIYENEGFRPYAEDTLSARGSSGDSDSPGSSSLSLTRKNSPKSGSPKSSSLLKLKAEKNAQAEMGKNHSSASFSSSITINTTCCSSSSSSSSSLSKTSGDLKPRSASDAGIRGTPKVRAKKDADANAGLTSCPRAKPSVRPKPFLNRAESQSQEKMDISTLRRQLRPTGQLRGGLKGSKSEDSELPPQTASEAPSEGSRRSSSDLITLPATTPPCPTKKEWEGPATSYMTCSAYQKVQDSEISFPAGVEVQVLEKQESGWWYVRFGELEGWAPSHYLVLDENEQPDPSGKELDTVPAKGRQNEGKSDSLEKIERRVQALNTVNQSKKATPPIPSKPPGGFGKTSGTPAVKMRNGVRQVAVRPQSVFVSPPPKDNNLSCALRRNESLTATDGLRGVRRNSSFSTARSAAAEAKGRLAERAASQGSDSPLLPAQRNSIPVSPVRPKPIEKSQFIHNNLKDVYVSIADYEGDEETAGFQEGVSMEVLERNPNGWWYCQILDGVKPFKGWVPSNYLEKKN
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9644
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.