missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35090-20ul
This item is not returnable.
View return policy
Description
SH3PX1 Polyclonal antibody specifically detects SH3PX1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SH3PX1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| MST155, MSTP155, Protein SDP1, SDP1, SH3 and PX domain-containing protein 1, SH3PX1WISP, SH3PXD3ASH3 and PX domain-containing protein SH3PX1, sorting nexin 9, sorting nexin-9, WiskSH3 and PX domain-containing protein 3A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human SH3PX1 (NP_057308.1).,, Sequence:, MATKARVMYDFAAEPGNNELTVNEGEIITITNPDVGGGWLEGRNIKGERGLVPTDYVEILPSDGKDQFSCGNSVADQAFLDSLSASTAQASSSAASNNHQVGSGNDPWSAWSASKSGNWESSEGWGAQPEGAGAQRNTNTPNNWDTAFGHPQAYQGPATGDDDDWDEDWDGPKSSSYFKDSESADAGGAQRGNSRASSSSMKIPLNKFPG | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51429 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction