missing translation for 'onlineSavingsMsg'
Learn More

SH3PX1 Antibody (3C6), Novus Biologicals™

Product Code. 18396809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396809 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396809 Supplier Novus Biologicals Supplier No. H00051429M0350ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SH3PX1 Monoclonal antibody specifically detects SH3PX1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3PX1
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 3C6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05022
Gene Alias MST155, MSTP155, Protein SDP1, SDP1, SH3 and PX domain-containing protein 1, SH3PX1WISP, SH3PXD3ASH3 and PX domain-containing protein SH3PX1, sorting nexin 9, sorting nexin-9, WiskSH3 and PX domain-containing protein 3A
Host Species Mouse
Immunogen SNX9 (AAH05022, 1 a.a. ~ 595 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MATKARVMYDFAAEPGNNELTVNEGEIITITNPDVGGGWLEGRNIKGERGLVPTDYVEILPSDGKDQFSCGNSVADQAFLDSLSASTAHASSSAASNNHQVGSGNDPWSAWSASKSGNWESSEGWGAQPEGAGAQRNTNTPNNWDTAFGHPQAYQGPATGDDDDWDEDWDGPKSSSYFKDSESADAGGAQRGNSRASSSSMKIPLNKFPGFAEPGTEQYLLAKQLAKPKEKIPIIVGDYGPMWVYPTSTFDCVVADPRKGSKMYGLKSYIEYQLTPTNTNRSVNHRYKHFDWLYERLLVKFGSAIPIPSLPDKQVTGRFEEEFIKMRMERLQAWMTRMCRHPVISESEVFQQFLNFRDEKEWKTGKRKAERDELAGVMIFSTMEPEAPDLDLVEIEQKCEAVGKFTKAMDDGVKELLTVGQEHWKRCTGPLPKEYQKIGKALQSLATVFSSSGYQGETDLNDAITEAGKTYEEIASLVAEQPKKDLHFLMECNHEYKGFLGCFPDVIGTHKGAIEKVKESDKLVATSKITLQDKQNMVKRVSIMSYALQAEMNHFHSNRIYDYNSVIRLYLEQQVQFYETIAEKLRQALSRFPVM
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51429
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.