missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3KBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570018
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Goat: 80%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
Specifications
| SH3KBP1 | |
| Polyclonal | |
| Unconjugated | |
| Sh3kbp1 | |
| 1200007H22Rik; 1700125L08Rik; 5830464D22Rik; AI447724; cbl-interacting protein of 85 kDa; c-Cbl-interacting protein; CD2-binding protein 3; CD2BP3; CIN85; GIG10; HSB1; HSB-1; Human Src family kinase-binding protein 1; IN85; LOW QUALITY PROTEIN: SH3 domain-containing kinase-binding protein 1; MIG18; migration-inducing gene 18; regulator of ubiquitous kinase; Ruk; Seta; SH3 domain containing kinase binding protein 1; SH3 domain-containing adapter protein; SH3 domain-containing kinase-binding protein 1; SH3-containing, expressed in tumorigenic astrocytes; SH3-domain kinase binding protein 1; Sh3kbp1; Src family kinase-binding protein 1; src-related kinase binding protein-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84357 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q925Q9 | |
| Sh3kbp1 | |
| synthetic peptide directed towards the C-terminal region of rat SH3KBP1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction