missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GLB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15897-1-AP-150UL
This item is not returnable.
View return policy
Description
Belongs to the endophilin family. 2 isoforms of the human protein are produced by alternative splicing. Pierrat B., Genomics 71:222-234(2001). Nagase T., DNA Res. 8:85-95(2001). Wan D., Proc. Natl. Acad. Sci. U. S. A. 101:15724-15729(2004).Specifications
| SH3GLB2 | |
| Polyclonal | |
| Unconjugated | |
| SH3GLB2 | |
| Endophilin B2, KIAA1848, PP6569, PP9455, SH3GLB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 227700, 311848, 56904 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PPJ9, Q8R3V5, Q9NR46 | |
| SH3GLB2 | |
| SH3GLB2 Fusion Protein Ag8712 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction