missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3GLB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106743
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SH3GLB2.
SH3GLB2 (SH3 Domain Containing GRB2 Like, Endophilin B2) is a Protein Coding gene. Among its related pathways are Endocytosis. An important paralog of this gene is SH3GLB1. [GeneCards].
Specifications
| SH3GLB2 | |
| Polyclonal | |
| Unconjugated | |
| SH3GLB2 | |
| EMBL:AAH87652.1}; endophilin-B2; Kiaa1848; PP578; PP6569; PP9455; retinoid receptor-induced gene 1; RRIG1; SH3 domain containing GRB2 like endophilin B2; SH3 domain-containing GRB2-like protein B2; SH3-containing protein SH3GLB2; SH3-domain GRB2-like endophilin B2; Sh3glb2; sh3glb2 {ECO:0000312; SPAS-1; stimulator of prostatic adenocarcinoma-specific T-cells-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 227700, 311848, 56904 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5PPJ9, Q8R3V5, Q9NR46 | |
| SH3GLB2 | |
| A synthesized peptide derived from human SH3GLB2(Accession Q9NR46), corresponding to amino acid residues A349-S395. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction