missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GLB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20346
This item is not returnable.
View return policy
Description
SH3GLB2 Polyclonal specifically detects SH3GLB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SH3GLB2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| endophilin-B2, GRB2-like, endophilin B2, SH3-containing protein SH3GLB2, SH3-domain GRB2-like endophilin B2 | |
| Rabbit | |
| 44 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.98mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9NR46 | |
| SH3GLB2 | |
| Recombinant protein encompassing a sequence within the center region of human SH3GLB2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 56904 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction