missing translation for 'onlineSavingsMsg'
Learn More

SH3GLB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97456-100ul

 View more versions of this product

Product Code. 18660262

  • £255.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SH3GLB2 Polyclonal antibody specifically detects SH3GLB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SH3GLB2
Polyclonal
Immunohistochemistry-Paraffin 1:50-1:200
endophilin-B2, GRB2-like, endophilin B2, SH3-containing protein SH3GLB2, SH3-domain GRB2-like endophilin B2
Produced in rabbits immunized with E. coli-derived Human SH3GLB2 fragment.
100 μg
Cancer, Endocrinology, Immunology
56904
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.