missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3GL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576161
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SH3GL3 is implicated in endocytosis. It may recruit other proteins to membranes with high curvature.
Specifications
| SH3GL3 | |
| Polyclonal | |
| Unconjugated | |
| Sh3gl3 | |
| CNSA3; EEN-B2; Endophilin 3; endophilin A3; endophilin III; endophilin-3; Endophilin-A3; HsT19371; SH3 domain containing GRB2 like 3; SH3 domain containing GRB2 like 3, endophilin A3; SH3 domain containing GRB2 like endophilin A3; SH3 domain protein 2 C1; SH3 domain protein 2 C2; SH3 domain protein 2C; SH3 domain-containing GRB2-like protein 3; Sh3d2c; Sh3d2c1; Sh3d2c2; SH3-domain GRB2-like 3; Sh3gl3; SH3p13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20408, 6457, 81921 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35180, Q62421, Q99963 | |
| Sh3gl3 | |
| Recombinant fusion protein of human SH3GL3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction