missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3GL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546726
This item is not returnable.
View return policy
Description
Peptide sequence: EAALDYHRQS TEILQELQSK LQMRISAASS VPRREYKPRP VKRSSSELNG.
SH3GL3 is implicated in endocytosis. It may recruit other proteins to membranes with high curvature.
Specifications
| SH3GL3 | |
| Polyclonal | |
| Unconjugated | |
| Sh3gl3 | |
| CNSA3; EEN-B2; Endophilin 3; endophilin A3; endophilin III; endophilin-3; Endophilin-A3; HsT19371; SH3 domain containing GRB2 like 3; SH3 domain containing GRB2 like 3, endophilin A3; SH3 domain containing GRB2 like endophilin A3; SH3 domain protein 2 C1; SH3 domain protein 2 C2; SH3 domain protein 2C; SH3 domain-containing GRB2-like protein 3; Sh3d2c; Sh3d2c1; Sh3d2c2; SH3-domain GRB2-like 3; Sh3gl3; SH3p13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6457 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99963 | |
| Sh3gl3 | |
| Synthetic peptide directed towards the middle region of human SH3GL3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction