missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3GL1 Monoclonal Antibody (GT3065)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538256
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| SH3GL1 | |
| Monoclonal | |
| 5.15 mg/mL | |
| PBS with no preservative | |
| O35964, Q62419, Q99961 | |
| SH3GL1 | |
| Recombinant protein encompassing a sequence within the center region of mouse SH3GL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT3065 | |
| Unconjugated | |
| SH3GL1 | |
| C77078; CNSA1; EEN; EEN fusion partner of MLL; endophilin A2; endophilin II; endophilin-2; Endophilin-A2; extra 11-19 leukemia fusion; extra eleven-nineteen leukemia fusion gene protein; MGC111371; SH3 domain containing GRB2 like 1; SH3 domain containing GRB2 like 1, endophilin A2; SH3 domain protein 2B; SH3 domain-containing GRB2-like protein 1; SH3-containing Grb-2-like 1 protein; Sh3d2b; SH3-domain GRB2-like 1; SH3GL1; Sh3p8 | |
| Mouse | |
| Protein A | |
| RUO | |
| 20405, 6455, 81922 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction