missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557992
This item is not returnable.
View return policy
Description
Immunogen sequence: EGTLIDLSEG FSETSFNDIK VPSPSALLVD NPTPFGNAKE VIAIKDYCPT NFTTLKFSKG DHLYVLDTSG GEWWYAHNT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
SH3BP4 was initially isolated as a novel cDNA using differential display analysis using cultured human corneal fibroblasts. The protein contains three Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. SH3BP4 has been shown to interact with specific endocytic proteins such as clathrin, dynamin, and the transferrin receptor (TfR) and localizes to TfR-containing coated pits and vesicles. It is thought to be involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of TfR. At least two isoforms of SH3BP4 are known to exist.
Specifications
| SH3BP4 | |
| Polyclonal | |
| Unconjugated | |
| SH3BP4 | |
| AI594717; AW227605; BOG25; EHB10; EH-binding protein 10; SH3 domain binding protein 4; SH3 domain-binding protein 4; SH3 domain-binding protein 4; SH3-domain binding protein 4; Sh3bp4; SH3-domain binding protein 4; si:dkey-57b6.3; Transferrin receptor-trafficking protein; TTP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P0V3 | |
| SH3BP4 | |
| Recombinant protein corresponding to Human SH3BP4. Recombinant protein control fragment (Product #RP-96078). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction