missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP4 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77031MFV450
This item is not returnable.
View return policy
Description
SH3BP4 Polyclonal antibody specifically detects SH3BP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SH3BP4 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| BOG25SH3 domain-binding protein 4, EHB10, EH-binding protein 10, SH3-domain binding protein 4, transferrin receptor trafficking protein, Transferrin receptor-trafficking protein, TTP | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human SH3BP4. The immunogen is located within amino acids 860 - 910 of SH3BP4. Amino Acid Squence: MDAYESPHRDRNGVVDSE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23677 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction