missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP2, Mouse anti-Human, Clone: 1E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006452-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: PLPNSVFVNTTESCEVERLFKATSPRGEPQDGLYCIRNSSTKSGKVLVVWDETSNKVRNYRIFEKDSKFYLEGEVLFVSVGSMVEHYHTHVLPSHQSLLLRHPYGYTGPRSpecifications
| SH3BP2 | |
| Monoclonal | |
| Unconjugated | |
| SH3-domain binding protein 2 | |
| 3BP2/CRBM/CRPM/FLJ42079/RES4-23 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1E9 | |
| In 1x PBS, pH 7.4 | |
| BC022996 | |
| SH3BP2 | |
| SH3BP2 (AAH22996, 452 a.a. ∼ 561 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6452 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction