missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SH3BP2 Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen SH3BP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067058
View Documents
Novus Biologicals™
NBP3-30802-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30066786
View Documents
Novus Biologicals™
NBP3-30802-250ug
250 μg
£386.00
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SH3BP2 Polyclonal antibody specifically detects SH3BP2 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

SH3BP2
Polyclonal
Purified
RUO
Mouse
Abl-SH3 binding protein 2, Cherubism, CRBM, CRPM, FLJ42079, FLJ54978,3BP-2,3BP2, RES4-23, SH3 domain-binding protein 2, SH3-domain binding protein 2, TNFAIP3 interacting protein 2
Antibody developed using SH2 domain of the SH3BP2 protein fused to GST.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
Unconjugated
Sheep
Cell Biology, Phospho Specific, Signal Transduction
Phosphate buffered saline
6452
IgG
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.