missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20541-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.Specifications
| SH3BP1 | |
| Polyclonal | |
| Unconjugated | |
| SH3BP1 | |
| 3BP 1, SH3 domain binding protein 1, SH3BP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20401, 23616 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55194, Q9Y3L3 | |
| Sh3bp1 | |
| SH3BP1 Fusion Protein Ag14575 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction