missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521851
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, Jurkat, NT2D1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.Specifications
SH3BP1 | |
Polyclonal | |
Unconjugated | |
SH3BP1 | |
3bp1; 3BP-1; ARHGAP43; OTTHUMP00000028981; SH3 domain binding protein 1; SH3 domain-binding protein 1; SH3BP1; SH3-domain binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y3L3 | |
SH3BP1 | |
Synthetic peptide corresponding to a region within amino acids 639 and 701 of SH3BP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |