missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BP1 Monoclonal Antibody (OTI1C9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527496
This item is not returnable.
View return policy
Description
SH3BP1 Monoclonal Antibody for Western Blot, IHC (P)
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.
Specifications
| SH3BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y3L3 | |
| SH3BP1 | |
| Full length human recombinant protein of SH3BP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C9 | |
| Unconjugated | |
| SH3BP1 | |
| 3bp1; 3BP-1; ARHGAP43; OTTHUMP00000028981; SH3 domain binding protein 1; SH3 domain-binding protein 1; SH3BP1; SH3-domain binding protein 1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23616 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction