missing translation for 'onlineSavingsMsg'
Få mere at vide

SH3BGRL3 Rabbit Polyclonal Antibody, Proteintech

Produktkode 30235702 Shop alle Proteintech produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
150 μL
20 μL
Förpackningsstorlek
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
30235702 150 μL 150µL
30236892 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 30235702 missing translation for 'mfr' Proteintech missing translation for 'supplierNo' 122101AP150UL

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Rabbit Polyclonal Antibody

The SH3BGRL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SH3BGRL3. This antibody recognizes human, mouse, rat antigen. The SH3BGRL3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.

Tekniske data

Antigen SH3BGRL3
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 900 μg/mL
Conjugate Unconjugated
Gene Symbols SH3BP-1; TIP-B1
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83442
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 10 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.