missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BGRL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556575
This item is not returnable.
View return policy
Description
Immunogen sequence: SEVTRILDGK RIQYQLVDIS QDNALRDEMR ALAGNPKATP PQIVNGDQYC GDYELFVEAV EQNTLQEFLK LA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.
SH3BGRL3 could act as a modulator of glutaredoxin biological activity.
Specifications
| SH3BGRL3 | |
| Polyclonal | |
| Unconjugated | |
| SH3BGRL3 | |
| 1110004L05Rik; AU022188; epididymis secretory protein Li 297; HEL-S-297; P1725; SH3 domain binding glutamate rich protein like 3; SH3 domain binding glutamate-rich protein like 3; SH3 domain binding glutamic acid-rich protein like 3; SH3 domain binding glutamic acid-rich protein-like 3; SH3 domain-binding glutamic acid-rich-like protein 3; SH3 domain-binding protein 1; SH3BGRL3; SH3BGRL3-like protein; SH3BP-1; TIP-B1; TNF inhibitory protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83442 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H299 | |
| SH3BGRL3 | |
| Recombinant protein corresponding to Human SH3BGRL3. Recombinant protein control fragment (Product #RP-94257). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction