missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BGRL Antibody (OTI8F7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | SH3BGRL |
|---|---|
| Clone | OTI8F7 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:150 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306422
|
Novus Biologicals
NBP3-45012-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306429
|
Novus Biologicals
NBP3-45012-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| SH3BGRL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MGC117402, SH3 domain binding glutamic acid-rich protein like, SH3 domain-binding glutamic acid-rich-like protein, SH3BGR, SH3-binding domain glutamic acid-rich protein like | |
| Full length human recombinant protein of human SH3BGRL (NP_003013) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI8F7 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 6451 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title