missing translation for 'onlineSavingsMsg'
Learn More

SH3BGRL Antibody (OTI2H5) [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30688766 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688766 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688766 Supplier Novus Biologicals Supplier No. NBP345014AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL
Applications Western Blot
Classification Monoclonal
Clone OTI2H5
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias MGC117402, SH3 domain binding glutamic acid-rich protein like, SH3 domain-binding glutamic acid-rich-like protein, SH3BGR, SH3-binding domain glutamic acid-rich protein like
Host Species Mouse
Immunogen Full length human recombinant protein of human SH3BGRL (NP_003013) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6451
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.