missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SH3BGRL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2H5 |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MGC117402, SH3 domain binding glutamic acid-rich protein like, SH3 domain-binding glutamic acid-rich-like protein, SH3BGR, SH3-binding domain glutamic acid-rich protein like |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human SH3BGRL (NP_003013) produced in E.coli. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?