missing translation for 'onlineSavingsMsg'
Learn More

SH3BGRL Antibody (OTI2C7), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen SH3BGRL
Clone OTI2C7
Dilution Western Blot 1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306595
View Documents
Novus Biologicals
NBP3-45013-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306741
View Documents
Novus Biologicals
NBP3-45013-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

SH3BGRL
Western Blot 1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
MGC117402, SH3 domain binding glutamic acid-rich protein like, SH3 domain-binding glutamic acid-rich-like protein, SH3BGR, SH3-binding domain glutamic acid-rich protein like
Full length human recombinant protein of human SH3BGRL(NP_003013) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI2C7
Western Blot
Unconjugated
Mouse
Signal Transduction
PBS (pH 7.3) 1% BSA, 50% glycerol
6451
IgG1
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.