missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BGR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569383
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Horse: 83%; Human: 100%.
SH3BGR is a protein coding gene.
Specifications
| SH3BGR | |
| Polyclonal | |
| Unconjugated | |
| Sh3bgr | |
| 21-GARP; 21-glutamic acid-rich protein; 5430437A18Rik; SH3 domain binding glutamate rich protein; SH3 domain binding glutamate-rich protein; SH3 domain binding glutamic acid-rich protein; SH3 domain-binding glutamic acid-rich protein; SH3BGR; SH3BGR protein; SH3-binding domain and glutamic acid-rich protein; SH3-binding domain glutamic acid-rich protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P55822 | |
| Sh3bgr | |
| synthetic peptide directed towards the N terminal region of human SH3BGR. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction