missing translation for 'onlineSavingsMsg'
Learn More

SH3BGR Antibody (3B7), Novus Biologicals™

Product Code. 18352789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352789 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352789 Supplier Novus Biologicals Supplier No. H00006450M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SH3BGR Monoclonal antibody specifically detects SH3BGR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGR
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3B7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06371
Gene Alias 21-GARP21-glutamic acid-rich protein, 21-Glutamic Acid-Rich Protein (21-GARP)10SH3-binding domain and glutamic acid-rich protein, SH3 domain binding glutamic acid-rich protein, SH3 domain-binding glutamic acid-rich protein, SH3BGR protein, SH3GBR
Host Species Mouse
Immunogen SH3BGR (AAH06371, 1 a.a. ~ 239 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPLLLLGETEPLKLERDCRSPVEPWAAASPDLALACLCHCQDLSSGAFPNRGVLGGVLFPTVEMVIKVFVATSSGSIAIRKKQQEVVGFLEANKIDFKELDIAGDEDNRRWMRENVPGEKKPQNGIPLPPQIFNEEQYCGDFDSFFSAKEENIIYSFLGLAPPPDSKGSEKAEEGGETEAQKEGSEDVGNLPEAQEKNEEEGETATEETEEIAMEGAEGEAEEEEETAEGEEPGEDEDS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6450
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.