missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH2D4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115892
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SH2D4A.
SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
Specifications
| SH2D4A | |
| Polyclonal | |
| Unconjugated | |
| SH2D4A | |
| 2210402M20Rik; FLJ20967; PPP1R38; protein phosphatase 1 regulatory subunit 38; protein phosphatase 1, regulatory subunit 38; protein SH(2)A; SH2 domain containing 4A; SH2 domain-containing 4A; SH2 domain-containing protein 4A; SH2A; SH2D4A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 63898 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9H788 | |
| SH2D4A | |
| A synthesized peptide derived from human SH2D4A(Accession Q9H788), corresponding to amino acid residues S177-K227. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction