missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D3C Monoclonal Antibody (1B6)

SH2D3C Monoclonal Antibody (1B6)
Brand: Invitrogen MA520896
This item is not returnable.
View return policy
Description
SH2D3C Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| SH2D3C | |
| Monoclonal | |
| 0.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8N5H7 | |
| SH2D3C | |
| SH2D3C (NP_005480, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 1B6 | |
| Unconjugated | |
| SH2D3C | |
| Cas/HEF1-associated signal transducer; CHAT; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10044 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction