missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH2D2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106742
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SH2D2A.
Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.
Specifications
| SH2D2A | |
| Polyclonal | |
| Unconjugated | |
| Sh2d2a | |
| F2771; Lad; Lck-associated adapter protein; p56Lck-associated adapter protein; Ribp; Rlk/Itk-binding protein; SCAP; SH2 domain containing 2A; SH2 domain protein 2A; SH2 domain-containing adapter protein; SH2 domain-containing protein 2A; SH22A; SH2D2A; T cell specific adapter protein TSAd; T cell-specific adapter protein; T lymphocyte specific adaptor protein; Tsad; VEGF receptor-associated protein; VRAP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9047 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NP31 | |
| Sh2d2a | |
| A synthesized peptide derived from human SH2D2A(Accession Q9NP31), corresponding to amino acid residues N303-P353. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction