missing translation for 'onlineSavingsMsg'
Learn More

SH2D1A Antibody (1C9), Novus Biologicals™

Product Code. 18335107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335107 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335107 Supplier Novus Biologicals Supplier No. H00004068M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SH2D1A Monoclonal antibody specifically detects SH2D1A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D1A
Applications Western Blot, Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone 1C9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS, pH 7.4
Gene Accession No. AAH20732
Gene Alias DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A
Host Species Mouse
Immunogen SH2D1A (AAH20732, 1 a.a. ~ 128 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDAVAVYHGKISRETGEKLLLATGLDGSYLLRDSESVPGVYCLCVLYHGYIYTYRVSQTETGSWSAETAPGVHKRYFRKIKNLISAFQKPDQGIVIPLQYPVEKKSSARSTQGTTGIREDPDVCLKAP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4068
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.