missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2B3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22198-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.Specifications
| SH2B3 | |
| Polyclonal | |
| Unconjugated | |
| SH2B3 | |
| LNK, Lymphocyte adapter protein, SH2B adapter protein 3, SH2B adaptor protein 3, SH2B3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10019 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UQQ2 | |
| SH2B3 | |
| SH2B3 Fusion Protein Ag17381 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction