missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22515
This item is not returnable.
View return policy
Description
SH2B2 Polyclonal specifically detects SH2B2 in Mouse samples. It is validated for Western Blot.
Specifications
| SH2B2 | |
| Polyclonal | |
| Western Blot 1:500 | |
| adaptor protein with pleckstrin homology and src homology 2 domains, APSAdapter protein with pleckstrin homology and Src homology 2 domains, SH2 and PH domain-containing adapter protein APS, SH2B adapter protein 2, SH2B adaptor protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10603 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS with 1 mg/ml BSA. with 0.05% Sodium Azide | |
| SH2B2 | |
| Synthetic peptide corresponding to residues E(605) A T L G R A R A V E N Q Y S(619) from mouse APS. | |
| 100 μg | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction