missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH2B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576759
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described.
Specifications
| SH2B1 | |
| Polyclonal | |
| Unconjugated | |
| SH2B1 | |
| AI425885; C530001K22Rik; DKFZp547G1110; fceRI gamma-chain-interacting protein SH2-B; FLJ30542; Irip; KIAA1299; mKIAA1299; pro-rich, PH and SH2 domain-containing signaling mediator; PSM; SH2 B; SH2 domain-containing protein 1B; SH2 domain-containing putative adapter SH2-B; Sh2b; SH2-B; SH2B adapter protein 1; SH2B adaptor protein 1; SH2-B PH domain containing signaling mediator 1; SH2-B PH domain-containing signaling mediator 1; SH2-B signaling protein; SH2B1; SH2B1 protein; SH2-Bb; Sh2bpsm1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20399, 25970, 89817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q62985, Q91ZM2, Q9NRF2 | |
| SH2B1 | |
| Recombinant full length Human SH2B1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction