missing translation for 'onlineSavingsMsg'
Learn More

SH2B1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30735769 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735769 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735769 Supplier Novus Biologicals Supplier No. AF6915Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

SH2B1 Polyclonal antibody specifically detects SH2B1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2B1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias FLJ30542, KIAA1299, Pro-rich, PH and SH2 domain-containing signaling mediator, PSM, SH2 domain-containing protein 1B, SH2 domain-containing putative adapter SH2-B, SH2B adapter protein 1, SH2B adaptor protein 1, SH2-B signaling protein, SH2BDKFZp547G1110
Host Species Goat
Immunogen E. coli-derived recombinant human SH2B1, Pro317-Leu467, Accession # Q9NRF2
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 25970
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.