missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGTA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115891
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SGTA.
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
Specifications
| SGTA | |
| Polyclonal | |
| Unconjugated | |
| SGTA | |
| 5330427H01Rik; AI194281; alpha h UBP; alphaSGT; Alpha-SGT; D10Ertd190e; hSGT; protein containing three tetratricopeptide repeats; Sgt; Sgt1; SGTA; small glutamine rich tetratricopeptide repeat containing alpha; small glutamine-rich protein with tetratricopeptide repeats 1; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR) containing protein (SGT); small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; small glutamine-rich tetratricopeptide repeat-containing protein alpha; Stg; UBP; vpu-binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 52551, 6449 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O43765, Q8BJU0 | |
| SGTA | |
| A synthesized peptide derived from human SGTA(Accession O43765), corresponding to amino acid residues A276-E313. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction