Learn More
SGTA Mouse anti-Human, CoraLite 594, Clone: 3B10B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60305-100UL
Description
The SGTA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SGTA. This antibody recognizes human, mouse antigen. The SGTA antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| SGTA | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Immunofluorescence | |
| 3B10B11 | |
| CoraLite 594 | |
| 100 μL | |
| 6449 | |
| IgG2b |