missing translation for 'onlineSavingsMsg'
Learn More

SGSM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97659-50ul

 View more versions of this product

Product Code. 18655284

  • £169.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SGSM3 Polyclonal antibody specifically detects SGSM3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SGSM3
Polyclonal
Immunocytochemistry/ Immunofluorescence 1:100-1:500
DJ1042K10.2, DKFZp761D051, MAP, merlin binding protein, Merlin-associated protein, rabGAPLP, Rab-GTPase-activating protein-like protein, RUN and SH3 containing 3, RUN and TBC1 domain containing 3, RUN and TBC1 domain-containing protein 3, RUSC3, RUTBC3, small G protein signaling modulator 3, small G protein signaling modulator 3 protein
Produced in rabbits immunized with E. coli-derived Human SGSM3 fragment.
50 μg
Signal Transduction
27352
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.