missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGSM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60726
This item is not returnable.
View return policy
Description
SGSM3 Polyclonal specifically detects SGSM3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| SGSM3 | |
| Polyclonal | |
| Western Blot 1:1000, Immunoprecipitation 8-15 ul/mg lysate | |
| DJ1042K10.2, DKFZp761D051, MAP, merlin binding protein, Merlin-associated protein, rabGAPLP, Rab-GTPase-activating protein-like protein, RUN and SH3 containing 3, RUN and TBC1 domain containing 3, RUN and TBC1 domain-containing protein 3, RUSC3, RUTBC3, small G protein signaling modulator 3, small G protein signaling modulator 3 protein | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
| SGSM3 | |
| between 699 and 749 | |
| 100 μL | |
| 27352 | |
| Store at 4degreesC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction