missing translation for 'onlineSavingsMsg'
Learn More

SGSM3 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18403306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403306 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403306 Supplier Novus Biologicals Supplier No. NBP281814AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SGSM3 Polyclonal antibody specifically detects SGSM3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DJ1042K10.2, DKFZp761D051, MAP, merlin binding protein, Merlin-associated protein, rabGAPLP, Rab-GTPase-activating protein-like protein, RUN and SH3 containing 3, RUN and TBC1 domain containing 3, RUN and TBC1 domain-containing protein 3, RUSC3, RUTBC3, small G protein signaling modulator 3, small G protein signaling modulator 3 protein
Host Species Rabbit
Immunogen SGSM3 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human SGSM3. The immunogen is located within amino acids 30 - 80 of SGSM3. Amino Acid Squence: YDEFGFRVYKEEGDEPGS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27352
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.