missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGSM2 Polyclonal Antibody
SGSM2 Polyclonal Antibody
Brand: Thermo Scientific PA554478
This item is not returnable.
View return policy
Description
SGSM2 Polyclonal Antibody for ICC/IF, IHC
Specifications
| SGSM2 | |
| Polyclonal | |
| Unconjugated | |
| SGSM2 | |
| D630003G22Rik; KIAA0397; mKIAA0397; R74628; RUN and TBC1 domain containing 1; RUN and TBC1 domain-containing protein 1; RUTBC1; Sgsm2; Small G protein signaling modulator 2; small G protein signaling modulator 2 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9905 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43147 | |
| SGSM2 | |
| Recombinant protein corresponding to Human SGSM2. Recombinant protein control fragment (Product #RP-93281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction