missing translation for 'onlineSavingsMsg'
Learn More

SGSM2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18371280 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371280 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371280 Supplier Novus Biologicals Supplier No. NBP282004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SGSM2 Polyclonal specifically detects SGSM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/mL
Formulation PBS
Gene Alias KIAA0397, RUN and TBC1 domain containing 1, RUN and TBC1 domain-containing protein 1, RUTBC1, small G protein signaling modulator 2, small G protein signaling modulator 2 protein
Gene Symbols SGSM2
Host Species Rabbit
Immunogen SGSM2 antibody was raised against an 18 amino acid synthetic peptide near the center of human SGSM2. The immunogen is located within amino acids 650 - 700 of SGSM2. Amino Acid Squence: VDDLEPPEPQDPEDSRPK
Molecular Weight of Antigen 116 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9905
Test Specificity Multiple isoforms of SGSM2 are known to exist. SGSM2 antibody is predicted to not cross react with other SGSM family proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.