missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGSH Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547426
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SGSH is one of several enzymes involved in the lysosomal degradation of heparan sulfate. Mutations in this gene are associated with Sanfilippo syndrome A, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. Transcripts of varying sizes have been reported but their biological validity has not been determined.
Specifications
| SGSH | |
| Polyclonal | |
| Unconjugated | |
| SGSH | |
| 4632406A19Rik; heparan sulfate sulfamidase; heparan sulfate sulfatase; HSS; MPS3A; mucopolysaccharidosis type IIIA; N-sulfoglucosamine sulfohydrolase; N-sulfoglucosamine sulfohydrolase (sulfamidase); N-sulphoglucosamine sulphohydrolase; SFMD; SGSH; Sulfoglucosamine sulfamidase; sulphamidase | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 27029 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| 0 | |
| SGSH | |
| Sf 21-derived recombinant mouse Sulfamidase/SGSH Arg23-Leu502. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction